Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMAN1
Entrez Gene ID
3998
Gene Name
lectin, mannose binding 1
Gene Aliases
ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58
Chromosome Location
Chr.18: 59327823 - 59359276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005570.3 NP_005561.1 6 - 7 794 80
GenBank mRNA AK223208.1 - 6 - 7 772 80
AK225108.1 - 6 - 7 772 80
AK225111.1 - 6 - 7 831 80
AK312869.1 - 6 - 7 772 80
BC017858.1 - 6 - 7 779 80
BC032330.1 - 6 - 7 777 80
U09716.1 - 6 - 7 782 80
X71661.1 - 6 - 7 783 80

View Full Product Details