The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080467.2 | NP_001073936.1 | 16 - 17 | 2290 | 85 |
GenBank mRNA | AB032945.2 | - | 16 - 17 | 2017 | 85 |
AB290160.1 | - | 16 - 17 | 2107 | 85 |