The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080467.2 | NP_001073936.1 | 27 - 28 | 3895 | 93 |
GenBank mRNA | AB032945.2 | - | 27 - 28 | 3625 | 93 |
AB290160.1 | - | 27 - 28 | 3715 | 93 | |
AK025336.1 | - | 7 - 8 | 1038 | 93 | |
AL133643.1 | - | 6 - 7 | 672 | 93 |