The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034837.2 | NP_001030009.1 | 3 - 4 | 761 | 69 |
NM_001034838.2 | NP_001030010.1 | 2 - 3 | 608 | 69 | |
NM_001278339.1 | NP_001265268.1 | 2 - 3 | 728 | 69 | |
NM_001278340.1 | NP_001265269.1 | 3 - 4 | 890 | 69 | |
NM_014592.3 | NP_055407.1 | 2 - 3 | 728 | 69 | |
XM_017009407.1 | XP_016864896.1 | 3 - 4 | 732 | 69 | |
GenBank mRNA | AF199597.1 | - | 2 - 3 | 191 | 69 |
AI884645.1 | - | 2 - 3 | 494 | 69 | |
AK074390.1 | - | 2 - 3 | 324 | 69 | |
AK301775.1 | - | 2 - 3 | 383 | 69 | |
AY170821.1 | - | 2 - 3 | 404 | 69 | |
AY780424.1 | - | 3 - 4 | 566 | 69 | |
BC035032.1 | - | 2 - 3 | 733 | 69 | |
BC050375.1 | - | 2 - 3 | 495 | 69 | |
DQ148476.1 | - | 2 - 3 | 218 | 69 | |
DQ148477.1 | - | 2 - 3 | 191 | 69 | |
DQ148478.1 | - | 3 - 4 | 224 | 69 | |
DQ148479.1 | - | 2 - 3 | 191 | 69 |