The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034837.2 | NP_001030009.1 | 8 - 9 | 1174 | 121 |
NM_001034838.2 | NP_001030010.1 | 7 - 8 | 1021 | 121 | |
NM_001278339.1 | NP_001265268.1 | 7 - 8 | 1216 | 121 | |
NM_001278340.1 | NP_001265269.1 | 8 - 9 | 1303 | 121 | |
NM_014592.3 | NP_055407.1 | 7 - 8 | 1141 | 121 | |
XM_017009407.1 | XP_016864896.1 | 8 - 9 | 1145 | 121 | |
GenBank mRNA | AF199597.1 | - | 7 - 8 | 604 | 121 |
AK074390.1 | - | 7 - 8 | 737 | 121 | |
AK301775.1 | - | 7 - 8 | 796 | 121 | |
AY170821.1 | - | 7 - 8 | 817 | 121 | |
AY780424.1 | - | 8 - 9 | 979 | 121 | |
BC035032.1 | - | 7 - 8 | 1146 | 121 | |
BC050375.1 | - | 7 - 8 | 908 | 121 | |
DQ148476.1 | - | 7 - 8 | 631 | 121 | |
DQ148477.1 | - | 7 - 8 | 604 | 121 | |
DQ148478.1 | - | 8 - 9 | 637 | 121 | |
DQ148479.1 | - | 7 - 8 | 679 | 121 |