Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CXCL14
- Entrez Gene ID
-
9547
- Gene Name
- C-X-C motif chemokine ligand 14
- Gene Aliases
-
BMAC, BRAK, KEC, KS1, MIP-2g, MIP2G, NJAC, SCYB14
- Chromosome Location
-
Chr.5: 135570679 - 135579279 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK056613.1 |
- |
1 - 1 |
228 |
94 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
R12978.1 |
- |
|
|
|
Chr. - - [-] on Build |
94 |
GenBankmRNA |
DA158249.1 |
- |
1 |
198 |
|
Chr. 5 - 135573203 - 135573768 [-] on Build GRCh38 |
94 |
GenBankmRNA |
DA761980.1 |
- |
1 |
228 |
|
Chr. 5 - 135573268 - 135573800 [-] on Build GRCh38 |
94 |
GenBankmRNA |
DA381392.1 |
- |
1 |
|
|
Chr. 5 - 135573300 - 135573932 [-] on Build GRCh38 |
94 |
GenBankmRNA |
R20080.1 |
- |
|
150 |
|
Chr. - - [-] on Build |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
neurogenin 1 |
NEUROG1 |
AKA, Math4C, NEUROD3, bHLHa6, ngn1 |
-37.3kb
|
Sense |
Chr. - 135534282 - 135535949 on Build GRCh38 |
Downstream |
uncharacterized LOC340074 |
LOC340074 |
|
74.9kb
|
Sense |
Chr. - 135648683 - 135653935 on Build GRCh38 |
View Full Product Details