The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005903.6 | NP_005894.3 | 1 - 2 | 122 | 151 |
GenBank mRNA | AF009745.1 | - | 1 - 2 | 117 | 148 |
AF071107.1 | - | 1 - 2 | 117 | 148 | |
AK313397.1 | - | 1 - 2 | 122 | 151 |