The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001419.2 | NP_001001419.1 | 6 - 7 | 1218 | 152 |
NM_001001420.2 | NP_001001420.1 | 4 - 5 | 1059 | 152 | |
NM_005903.6 | NP_005894.3 | 5 - 6 | 1134 | 152 | |
XM_017009470.1 | XP_016864959.1 | 6 - 7 | 1350 | 152 | |
GenBank mRNA | AF009678.1 | - | 3 - 4 | 786 | 152 |
AF010601.1 | - | 3 - 4 | 903 | 152 | |
AK313397.1 | - | 5 - 6 | 1134 | 152 | |
BC009682.2 | - | 3 - 4 | 940 | 152 | |
CR457424.1 | - | 3 - 4 | 772 | 152 | |
CR749366.1 | - | 5 - 6 | 1073 | 152 | |
CR749473.1 | - | 5 - 6 | 1061 | 152 | |
U59913.1 | - | 3 - 4 | 902 | 152 | |
U73825.1 | - | 3 - 4 | 921 | 152 |