The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000828.4 | NP_000819.3 | 12 - 13 | 2374 | 86 |
NM_007325.4 | NP_015564.4 | 12 - 13 | 2374 | 86 | |
GenBank mRNA | BC117464.1 | - | 12 - 13 | 2246 | 86 |
U10301.1 | - | 12 - 13 | 2134 | 86 | |
U10302.1 | - | 12 - 13 | 2134 | 86 |