The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166136.1 | NP_001159608.1 | 13 - 14 | 2545 | 151 |
NM_147127.4 | NP_667338.3 | 13 - 14 | 2107 | 151 | |
XM_017007735.1 | XP_016863224.1 | 13 - 14 | 2118 | 151 | |
XM_017007736.1 | XP_016863225.1 | 14 - 15 | 2096 | 151 | |
XM_017007737.1 | XP_016863226.1 | 14 - 15 | 1983 | 151 | |
XM_017007738.1 | XP_016863227.1 | 13 - 14 | 2118 | 151 | |
XM_017007739.1 | XP_016863228.1 | 14 - 15 | 2141 | 151 | |
GenBank mRNA | AB083067.1 | - | 12 - 13 | 1813 | 151 |
AY152402.1 | - | 13 - 14 | 2542 | 151 | |
AY152403.1 | - | 13 - 14 | 2542 | 151 | |
AY185210.1 | - | 13 - 14 | 2107 | 151 | |
BC143836.1 | - | 14 - 15 | 2146 | 151 |