The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166136.1 | NP_001159608.1 | 9 - 10 | 1644 | 64 |
NM_147127.4 | NP_667338.3 | 9 - 10 | 1206 | 64 | |
XM_017007735.1 | XP_016863224.1 | 9 - 10 | 1217 | 64 | |
XM_017007736.1 | XP_016863225.1 | 10 - 11 | 1195 | 64 | |
XM_017007737.1 | XP_016863226.1 | 10 - 11 | 1082 | 64 | |
XM_017007738.1 | XP_016863227.1 | 9 - 10 | 1217 | 64 | |
XM_017007739.1 | XP_016863228.1 | 10 - 11 | 1248 | 64 | |
GenBank mRNA | AB083067.1 | - | 8 - 9 | 912 | 64 |
AY152402.1 | - | 9 - 10 | 1641 | 64 | |
AY152403.1 | - | 9 - 10 | 1641 | 64 | |
AY185210.1 | - | 9 - 10 | 1206 | 64 | |
BC143836.1 | - | 10 - 11 | 1253 | 64 |