The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300860.1 | NP_001287789.1 | 11 - 12 | 1669 | 89 |
NM_014829.3 | NP_055644.2 | 11 - 12 | 1669 | 89 | |
NR_125341.1 | 10 - 11 | 1525 | 89 | ||
XM_011543758.1 | XP_011542060.1 | 11 - 12 | 1632 | 89 | |
GenBank mRNA | AB018344.1 | - | 11 - 12 | 1608 | 89 |
AF106680.1 | - | 11 - 12 | 1609 | 89 | |
AK126011.1 | - | 10 - 11 | 1480 | 89 | |
AK291788.1 | - | 11 - 12 | 1622 | 89 | |
BC012304.2 | - | 11 - 12 | 1599 | 89 |