The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278616.1 | NP_001265545.1 | 2 - 3 | 278 | 97 |
NM_001278617.1 | NP_001265546.1 | 3 - 4 | 338 | 97 | |
NM_004336.4 | NP_004327.1 | 3 - 4 | 338 | 97 | |
GenBank mRNA | AF043294.2 | - | 3 - 4 | 258 | 97 |
AF046078.1 | - | 3 - 4 | 275 | 97 | |
AF047471.1 | - | 3 - 4 | 261 | 97 | |
AK023540.1 | - | 3 - 4 | 294 | 97 | |
AK225317.1 | - | 3 - 4 | 294 | 97 | |
AK291221.1 | - | 3 - 4 | 291 | 97 | |
AK302418.1 | - | 2 - 3 | 234 | 97 | |
BC017038.1 | - | 3 - 4 | 259 | 97 | |
BC028201.1 | - | 3 - 4 | 259 | 97 | |
BP313176.1 | - | 3 - 4 | 318 | 97 | |
DB453210.1 | - | 3 - 4 | 343 | 97 |