Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR88
Entrez Gene ID
126248
Gene Name
WD repeat domain 88
Gene Aliases
PQWD
Chromosome Location
Chr.19: 33132092 - 33175799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173479.3 NP_775750.3 9 - 10 1222 155
NR_135565.1 9 - 10 1222 155
XM_011526449.2 XP_011524751.1 9 - 10 1216 155
GenBank mRNA AK131444.1 - 9 - 10 1581 155
BC031227.1 - 9 - 10 1222 155

View Full Product Details