Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP47
Entrez Gene ID
286464
Gene Name
cilia and flagella associated protein 47
Gene Aliases
CHDC2, CXorf22, CXorf30, CXorf59
Chromosome Location
Chr.X: 35919734 - 36385319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304548.1 NP_001291477.1 15 - 16 2784 89
NM_152632.3 NP_689845.2 15 - 16 2784 89
XM_017029452.1 XP_016884941.1 15 - 16 2777 89
XM_017029453.1 XP_016884942.1 15 - 16 2777 89
GenBank mRNA BC027936.1 - 15 - 16 2639 89
DV080632.1 - 2 - 3 89

View Full Product Details