The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143836.2 | NP_001137308.1 | 14 - 15 | 1648 | 87 |
NM_001143837.1 | NP_001137309.1 | 18 - 19 | 1913 | 87 | |
NM_001291926.1 | NP_001278855.1 | 15 - 16 | 1764 | 87 | |
NM_001291927.1 | NP_001278856.1 | 15 - 16 | 1518 | 87 | |
NM_001291929.1 | NP_001278858.1 | 14 - 15 | 1431 | 87 | |
NM_001300995.1 | NP_001287924.1 | 14 - 15 | 1371 | 87 | |
NM_016931.4 | NP_058627.1 | 15 - 16 | 1768 | 87 | |
XM_006718849.3 | XP_006718912.1 | 14 - 15 | 1576 | 87 | |
XM_011542857.2 | XP_011541159.1 | 15 - 16 | 1568 | 87 | |
XM_017017841.1 | XP_016873330.1 | 14 - 15 | 1615 | 87 | |
XM_017017842.1 | XP_016873331.1 | 11 - 12 | 1067 | 87 | |
XM_017017844.1 | XP_016873333.1 | 16 - 17 | 1754 | 87 | |
XM_017017845.1 | XP_016873334.1 | 12 - 13 | 1170 | 87 | |
GenBank mRNA | AB041035.1 | - | 15 - 16 | 1688 | 87 |
AF254621.1 | - | 15 - 16 | 1535 | 87 | |
AF261943.1 | - | 15 - 16 | 1449 | 87 | |
AJ704725.1 | - | 15 - 16 | 1445 | 87 | |
AJ704726.1 | - | 14 - 15 | 1329 | 87 | |
AJ704727.1 | - | 12 - 13 | 1004 | 87 | |
AJ704728.1 | - | 6 - 7 | 528 | 87 | |
AJ704729.1 | - | 5 - 6 | 408 | 87 | |
AK057189.1 | - | 18 - 19 | 1913 | 87 | |
AK291830.1 | - | 14 - 15 | 1411 | 87 | |
AK298323.1 | - | 14 - 15 | 1342 | 87 | |
AK298328.1 | - | 15 - 16 | 1518 | 87 | |
AK298357.1 | - | 14 - 15 | 1422 | 87 | |
AY288918.1 | - | 14 - 15 | 1329 | 87 | |
BC040105.1 | - | 15 - 16 | 1521 | 87 |