The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185059.1 | NP_001171988.1 | 13 - 14 | 3016 | 112 |
NM_024617.3 | NP_078893.2 | 13 - 14 | 3126 | 112 | |
XM_005252207.3 | XP_005252264.1 | 13 - 14 | 3078 | 112 | |
XM_005252208.2 | XP_005252265.1 | 13 - 14 | 3078 | 112 | |
XM_006717283.1 | XP_006717346.1 | 13 - 14 | 3078 | 112 | |
XM_011519011.2 | XP_011517313.1 | 13 - 14 | 3078 | 112 | |
XM_011519012.2 | XP_011517314.1 | 13 - 14 | 3026 | 112 | |
XM_011519013.2 | XP_011517315.1 | 13 - 14 | 3695 | 112 | |
XM_011519014.2 | XP_011517316.1 | 13 - 14 | 3297 | 112 | |
XM_011519015.1 | XP_011517317.1 | 13 - 14 | 3078 | 112 | |
XM_011519016.1 | XP_011517318.1 | 13 - 14 | 3078 | 112 | |
XM_017015130.1 | XP_016870619.1 | 13 - 14 | 3694 | 112 | |
XM_017015131.1 | XP_016870620.1 | 13 - 14 | 3336 | 112 | |
XM_017015132.1 | XP_016870621.1 | 14 - 15 | 3241 | 112 | |
GenBank mRNA | AB051498.1 | - | 5 - 6 | 1664 | 112 |
AK055948.1 | - | 1 - 2 | 976 | 112 | |
AL832193.1 | - | 14 - 15 | 3168 | 112 | |
BC144523.1 | - | 12 - 13 | 2914 | 112 | |
BX641077.1 | - | 12 - 13 | 4476 | 112 | |
CR933643.1 | - | 13 - 14 | 2990 | 112 | |
CR936608.1 | - | 14 - 15 | 3171 | 112 |