The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185074.1 | NP_001172003.1 | 1 - 2 | 219 | 100 |
NM_024617.3 | NP_078893.2 | 1 - 2 | 219 | 100 | |
XM_005252207.3 | XP_005252264.1 | 1 - 2 | 171 | 100 | |
XM_005252208.2 | XP_005252265.1 | 1 - 2 | 171 | 100 | |
XM_006717283.1 | XP_006717346.1 | 1 - 2 | 171 | 100 | |
XM_011519011.2 | XP_011517313.1 | 1 - 2 | 171 | 100 | |
XM_011519015.1 | XP_011517317.1 | 1 - 2 | 171 | 100 | |
XM_011519016.1 | XP_011517318.1 | 1 - 2 | 171 | 100 | |
XM_017015132.1 | XP_016870621.1 | 1 - 2 | 171 | 100 | |
GenBank mRNA | AK055546.1 | - | 1 - 2 | 144 | 100 |
AK308352.1 | - | 1 - 2 | 74 | 100 | |
AL832026.1 | - | 1 - 2 | 186 | 100 | |
AL832193.1 | - | 1 - 2 | 143 | 100 | |
BC032456.1 | - | 1 - 2 | 126 | 100 | |
BX641077.1 | - | 1 - 2 | 195 | 100 | |
CR933644.1 | - | 1 - 2 | 146 | 100 | |
CR936608.1 | - | 1 - 2 | 143 | 100 | |
DC377509.1 | - | 1 - 2 | 219 | 100 |