The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040462.2 | NP_001035552.1 | 6 - 7 | 1064 | 111 |
NM_001159707.1 | NP_001153179.1 | 5 - 6 | 716 | 111 | |
NM_001159708.1 | NP_001153180.1 | 5 - 6 | 1173 | 220 | |
NM_001159709.1 | NP_001153181.1 | 6 - 7 | 767 | 111 | |
NM_001159710.1 | NP_001153182.1 | 5 - 6 | 567 | 111 | |
NM_024850.2 | NP_079126.1 | 6 - 7 | 1194 | 241 | |
XM_011534650.2 | XP_011532952.1 | 6 - 7 | 869 | 111 | |
XM_011534651.2 | XP_011532953.1 | 5 - 6 | 1883 | 111 | |
XM_011534652.2 | XP_011532954.1 | 5 - 6 | 1859 | 111 | |
XM_011534653.2 | XP_011532955.1 | 4 - 5 | 420 | 111 | |
GenBank mRNA | AK025111.1 | - | 6 - 7 | 1112 | 241 |
AK225213.1 | - | 6 - 7 | 1112 | 241 | |
AK296597.1 | - | 6 - 7 | 722 | 111 | |
AK298687.1 | - | 5 - 6 | 567 | 111 | |
AY358523.1 | - | 6 - 7 | 1012 | 111 | |
BC119696.1 | - | 5 - 6 | 519 | 111 |