The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004246.2 | NP_004237.1 | 1 - 2 | 698 | 81 |
XM_011524077.2 | XP_011522379.1 | 1 - 2 | 737 | 81 | |
XM_017025339.1 | XP_016880828.1 | 1 - 2 | 737 | 81 | |
GenBank mRNA | AF105367.1 | - | 1 - 2 | 185 | 81 |
AK307283.1 | - | 1 - 2 | 345 | 81 | |
AK314269.1 | - | 1 - 2 | 698 | 81 | |
BC096261.1 | - | 1 - 2 | 185 | 81 | |
BC096262.1 | - | 1 - 2 | 185 | 81 | |
BC096263.1 | - | 1 - 2 | 181 | 81 | |
BC096264.1 | - | 1 - 2 | 185 | 81 |