The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004246.2 | NP_004237.1 | 4 - 5 | 1019 | 68 |
XM_011524077.2 | XP_011522379.1 | 5 - 6 | 1133 | 68 | |
XM_017025339.1 | XP_016880828.1 | 4 - 5 | 1076 | 68 | |
GenBank mRNA | AK295765.1 | - | 4 - 5 | 398 | 68 |
AK314269.1 | - | 4 - 5 | 1019 | 68 | |
BC019818.1 | - | 1 - 2 | 64 | 68 | |
BC041176.1 | - | 1 - 2 | 75 | 68 | |
BC043390.1 | - | 2 - 3 | 214 | 68 | |
BC096261.1 | - | 4 - 5 | 506 | 68 | |
BC096262.1 | - | 4 - 5 | 506 | 68 | |
BC096263.1 | - | 4 - 5 | 502 | 68 | |
BC096264.1 | - | 4 - 5 | 506 | 68 |