The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300867.1 | NP_001287796.1 | 18 - 19 | 1780 | 100 |
NM_001324062.1 | NP_001310991.1 | 23 - 24 | 4982 | 100 | |
NM_001324063.1 | NP_001310992.1 | 21 - 22 | 4805 | 100 | |
NM_001324064.1 | NP_001310993.1 | 24 - 25 | 5027 | 100 | |
NM_001324065.1 | NP_001310994.1 | 18 - 19 | 1780 | 100 | |
NM_198551.3 | NP_940953.2 | 23 - 24 | 4982 | 100 | |
XM_006711304.3 | XP_006711367.2 | 21 - 22 | 5117 | 100 | |
XM_017001243.1 | XP_016856732.1 | 23 - 24 | 4807 | 100 | |
GenBank mRNA | AK025122.1 | - | 15 - 16 | 1281 | 100 |
AK025506.1 | - | 20 - 21 | 2662 | 100 | |
AK290335.1 | - | 14 - 15 | 1257 | 100 | |
AK296712.1 | - | 18 - 19 | 1761 | 100 | |
AX775845.1 | - | 18 - 19 | 1754 | 100 | |
BP423646.1 | - | 1 - 2 | 47 | 100 | |
D87742.1 | - | 20 - 21 | 2831 | 100 |