Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TRPM3
- Entrez Gene ID
-
80036
- Gene Name
- transient receptor potential cation channel subfamily M member 3
- Gene Aliases
-
GON-2, LTRPC3, MLSN2
- Chromosome Location
-
Chr.9: 70529063 - 71446989 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK021788.1 |
- |
6 - 7 |
853 |
83 |
View Full Product Details