The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000715.3 | NP_000706.1 | 6 - 7 | 907 | 74 |
XM_005273251.1 | XP_005273308.1 | 6 - 7 | 863 | 74 | |
XM_005273252.4 | XP_005273309.1 | 6 - 7 | 1039 | 74 | |
GenBank mRNA | AK303793.1 | - | 7 - 8 | 718 | 74 |
AK303798.1 | - | 6 - 7 | 857 | 74 | |
AK313164.1 | - | 6 - 7 | 853 | 74 | |
BC022312.1 | - | 6 - 7 | 907 | 74 | |
M31452.1 | - | 6 - 7 | 851 | 74 | |
X02865.1 | - | 4 - 5 | 476 | 74 |