The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032019.5 | NP_114408.3 | 8 - 9 | 1126 | 96 |
GenBank mRNA | AF393962.1 | - | 8 - 9 | 753 | 96 |
AF407272.1 | - | 8 - 9 | 753 | 96 | |
AF426160.1 | - | 8 - 9 | 1106 | 96 | |
BC032049.1 | - | 8 - 9 | 939 | 96 | |
BC094734.1 | - | 7 - 8 | 901 | 96 | |
BC125083.1 | - | 8 - 9 | 800 | 96 | |
CR456465.1 | - | 8 - 9 | 809 | 96 |