The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020928.1 | NP_065979.1 | 11 - 12 | 2542 | 110 |
XM_017009677.1 | XP_016865166.1 | 9 - 10 | 1493 | 110 | |
GenBank mRNA | AB046797.1 | - | 7 - 8 | 1128 | 110 |