The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020928.1 | NP_065979.1 | 4 - 5 | 1332 | 100 |
XM_017009677.1 | XP_016865166.1 | 2 - 3 | 283 | 100 |