Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX12
Entrez Gene ID
5193
Gene Name
peroxisomal biogenesis factor 12
Gene Aliases
PAF-3, PBD3A
Chromosome Location
Chr.17: 35574795 - 35578637 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000286.2 NP_000277.1 1 - 2 740 64
GenBank mRNA AB004546.1 - 1 - 2 309 64
AK312635.1 - 1 - 2 653 64
BC015751.1 - 1 - 2 655 64
BC031085.1 - 1 - 2 644 64
U91521.1 - 1 - 2 671 64

View Full Product Details