Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMBRA1
Entrez Gene ID
55626
Gene Name
autophagy and beclin 1 regulator 1
Gene Aliases
DCAF3, WDR94
Chromosome Location
Chr.11: 46396412 - 46594069 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267782.1 NP_001254711.1 18 - 19 3574 79
NM_001267783.1 NP_001254712.1 16 - 17 3208 79
NM_001300731.1 NP_001287660.1 15 - 16 3385 79
NM_017749.3 NP_060219.2 17 - 18 3293 79
XM_005253009.3 XP_005253066.1 16 - 17 3561 79
XM_005253011.3 XP_005253068.1 15 - 16 3474 79
XM_005253014.3 XP_005253071.1 17 - 18 3291 79
XM_006718259.2 XP_006718322.1 16 - 17 3442 79
XM_006718260.2 XP_006718323.1 15 - 16 3024 79
XM_017018007.1 XP_016873496.1 14 - 15 3294 79
GenBank mRNA AB051523.1 - 15 - 16 3337 79
AK000301.1 - 18 - 19 3524 79
AK023197.1 - 10 - 11 1816 79
AK302808.1 - 16 - 17 3623 79
AL834190.1 - 16 - 17 3088 79
BC045609.1 - 17 - 18 3293 79
DQ870924.1 - 14 - 15 3116 79

View Full Product Details