The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017919.1 | NP_001017919.1 | 5 - 6 | 980 | 80 |
NM_033544.2 | NP_291022.2 | 6 - 7 | 1064 | 80 | |
GenBank mRNA | BC064606.1 | - | 3 - 4 | 991 | 80 |
BC094739.1 | - | 5 - 6 | 1267 | 80 | |
BC113826.1 | - | 3 - 4 | 388 | 80 | |
BC140708.1 | - | 5 - 6 | 978 | 80 | |
BC140709.1 | - | 5 - 6 | 978 | 80 |