Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USH1C
Entrez Gene ID
10083
Gene Name
USH1 protein network component harmonin
Gene Aliases
AIE-75, DFNB18, DFNB18A, NY-CO-37, NY-CO-38, PDZ-45, PDZ-73, PDZ-73/NY-CO-38, PDZ73, PDZD7C, ush1cpst
Chromosome Location
Chr.11: 17493895 - 17544416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297764.1 NP_001284693.1 11 - 12 1065 91
NM_005709.3 NP_005700.2 12 - 13 1122 91
NM_153676.3 NP_710142.1 12 - 13 1122 91
NR_123738.1 12 - 13 1122 91
XM_011519832.2 XP_011518134.1 12 - 13 1122 91
XM_011519834.2 XP_011518136.1 12 - 13 1122 91
XM_017017072.1 XP_016872561.1 12 - 13 1122 91
XM_017017073.1 XP_016872562.1 11 - 12 1065 91
XM_017017074.1 XP_016872563.1 12 - 13 1122 91
XM_017017075.1 XP_016872564.1 12 - 13 1122 91
GenBank mRNA AB006955.1 - 11 - 12 1165 91
AB018687.1 - 12 - 13 1102 91
AF039699.1 - 12 - 13 1109 91
AF039700.1 - 12 - 13 1109 91
AK225614.1 - 12 - 13 1122 91
AK290788.1 - 12 - 13 1074 91
AK300936.1 - 12 - 13 881 91
BC016057.1 - 11 - 12 1049 91
KU178498.1 - 11 - 12 956 91
KU178499.1 - 12 - 13 1013 91

View Full Product Details