The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297764.1 | NP_001284693.1 | 4 - 5 | 497 | 66 |
NM_005709.3 | NP_005700.2 | 4 - 5 | 497 | 66 | |
NM_153676.3 | NP_710142.1 | 4 - 5 | 497 | 66 | |
NR_123738.1 | 4 - 5 | 497 | 66 | ||
XM_011519832.2 | XP_011518134.1 | 4 - 5 | 497 | 66 | |
XM_011519834.2 | XP_011518136.1 | 4 - 5 | 497 | 66 | |
XM_017017072.1 | XP_016872561.1 | 4 - 5 | 497 | 66 | |
XM_017017073.1 | XP_016872562.1 | 4 - 5 | 497 | 66 | |
XM_017017074.1 | XP_016872563.1 | 4 - 5 | 497 | 66 | |
XM_017017075.1 | XP_016872564.1 | 4 - 5 | 497 | 66 | |
GenBank mRNA | AB006955.1 | - | 3 - 4 | 540 | 66 |
AB018687.1 | - | 4 - 5 | 477 | 66 | |
AF039699.1 | - | 4 - 5 | 484 | 66 | |
AF039700.1 | - | 4 - 5 | 484 | 66 | |
AK225614.1 | - | 4 - 5 | 497 | 66 | |
AK290788.1 | - | 4 - 5 | 449 | 66 | |
AK300936.1 | - | 4 - 5 | 256 | 66 | |
BC016057.1 | - | 4 - 5 | 481 | 66 | |
KU178498.1 | - | 4 - 5 | 388 | 66 | |
KU178499.1 | - | 4 - 5 | 388 | 66 |