The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144666.2 | NP_653267.2 | 7 - 8 | 1960 | 65 |
NM_173589.3 | NP_775860.3 | 6 - 7 | 2063 | 65 | |
GenBank mRNA | AK093028.1 | - | 3 - 4 | 1144 | 65 |
BC111765.1 | - | 5 - 6 | 1813 | 65 | |
BX648742.1 | - | 6 - 7 | 2058 | 65 |