The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160364.2 | NP_001153836.1 | 11 - 12 | 1783 | 61 |
NM_001300732.1 | NP_001287661.1 | 11 - 12 | 1685 | 61 | |
NM_001324263.1 | NP_001311192.1 | 11 - 12 | 1672 | 61 | |
NM_001324265.1 | NP_001311194.1 | 11 - 12 | 1685 | 61 | |
NM_001324268.1 | NP_001311197.1 | 10 - 11 | 1537 | 61 | |
NM_024847.4 | NP_079123.3 | 11 - 12 | 1685 | 61 | |
NR_136733.1 | 11 - 12 | 1672 | 61 | ||
GenBank mRNA | AK024893.1 | - | 7 - 8 | 894 | 61 |
AK094664.1 | - | 11 - 12 | 1685 | 61 | |
AY236498.1 | - | 11 - 12 | 1619 | 61 | |
AY263165.1 | - | 11 - 12 | 1614 | 61 | |
BC036205.1 | - | 11 - 12 | 1614 | 61 | |
BC047719.1 | - | 11 - 12 | 1616 | 61 | |
CR933669.1 | - | 11 - 12 | 1783 | 61 |