Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTK2B
Entrez Gene ID
2185
Gene Name
protein tyrosine kinase 2 beta
Gene Aliases
CADTK, CAKB, FADK2, FAK2, PKB, PTK, PYK2, RAFTK
Chromosome Location
Chr.8: 27311482 - 27459391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004103.4 NP_004094.3 13 - 14 1399 75
NM_173174.2 NP_775266.1 17 - 18 1954 75
NM_173175.2 NP_775267.1 12 - 13 1301 75
NM_173176.2 NP_775268.1 12 - 13 1386 75
XM_005273447.4 XP_005273504.1 12 - 13 1314 75
XM_005273448.4 XP_005273505.1 12 - 13 1363 75
XM_011544441.2 XP_011542743.1 12 - 13 1324 75
XM_011544442.2 XP_011542744.1 12 - 13 1324 75
XM_017013214.1 XP_016868703.1 13 - 14 1389 75
XM_017013215.1 XP_016868704.1 12 - 13 1285 75
XM_017013216.1 XP_016868705.1 12 - 13 1324 75
GenBank mRNA AB209085.1 - 12 - 13 1296 75
AK128371.1 - 4 - 5 732 75
AY429564.1 - 5 - 6 427 75
BC036651.2 - 17 - 18 1954 75
BC042599.2 - 12 - 13 1331 75
L49207.1 - 12 - 13 1326 75
U33284.1 - 12 - 13 1253 75
U43522.1 - 13 - 14 1389 75

View Full Product Details