Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS6
Entrez Gene ID
203286
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Gene Aliases
ANKRD14, NPHP16, PKDR1, SAMD6
Chromosome Location
Chr.9: 98732009 - 98796542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173551.4 NP_775822.3 4 - 5 1160 69
XM_005251793.4 XP_005251850.1 4 - 5 1147 69
XM_005251794.4 XP_005251851.1 4 - 5 1147 69
XM_006716998.3 XP_006717061.1 4 - 5 1147 69
XM_006716999.2 XP_006717062.1 4 - 5 2232 69
XM_011518357.2 XP_011516659.1 4 - 5 3229 69
XM_017014445.1 XP_016869934.1 4 - 5 1147 69
XM_017014446.1 XP_016869935.1 3 - 4 3108 69
GenBank mRNA AK126614.1 - 2 - 3 214 69
AK127702.1 - 3 - 4 747 69
BC064367.1 - 1 - 2 147 69
CR749467.1 - 4 - 5 831 69
CR749472.1 - 2 - 3 511 69

View Full Product Details