Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS6
Entrez Gene ID
203286
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Gene Aliases
ANKRD14, NPHP16, PKDR1, SAMD6
Chromosome Location
Chr.9: 98732009 - 98796542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173551.4 NP_775822.3 10 - 11 2021 117
XM_005251793.4 XP_005251850.1 10 - 11 2008 117
XM_005251794.4 XP_005251851.1 10 - 11 2008 117
XM_006716998.3 XP_006717061.1 10 - 11 2008 117
XM_006716999.2 XP_006717062.1 10 - 11 3093 117
XM_011518357.2 XP_011516659.1 10 - 11 4090 117
XM_017014445.1 XP_016869934.1 10 - 11 2008 117
XM_017014446.1 XP_016869935.1 9 - 10 3969 117
XM_017014447.1 XP_016869936.1 6 - 7 802 117
GenBank mRNA AK094247.1 - 3 - 4 375 117
AK126614.1 - 8 - 9 1075 117
AK127702.1 - 9 - 10 1608 117
BC064367.1 - 7 - 8 1008 117
CR749467.1 - 10 - 11 1692 117
CR749472.1 - 8 - 9 1372 117

View Full Product Details