Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC11
Entrez Gene ID
114132
Gene Name
sialic acid binding Ig like lectin 11
Gene Aliases
-
Chromosome Location
Chr.19: 49948993 - 49961172 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052884.2 NP_443116.2 8 - 9 1738 149
XM_005258476.3 XP_005258533.2 7 - 8 1631 149
GenBank mRNA AB072038.1 - 8 - 9 1657 149
AF337818.1 - 8 - 9 1738 149

View Full Product Details