Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTPD4
Entrez Gene ID
9583
Gene Name
ectonucleoside triphosphate diphosphohydrolase 4
Gene Aliases
LALP70, LAP70, LYSAL1, NTPDase-4, UDPase
Chromosome Location
Chr.8: 23429152 - 23457731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128930.2 NP_001122402.1 8 - 9 1132 103
GenBank mRNA AB002390.1 - 6 - 7 686 103
AF016032.1 - 8 - 9 1061 103
BC034477.2 - 8 - 9 1096 103

View Full Product Details