Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLUL1
Entrez Gene ID
27098
Gene Name
clusterin like 1
Gene Aliases
RA337M
Chromosome Location
Chr.18: 596998 - 650298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289036.1 NP_001275965.1 4 - 5 495 87
NM_001318522.1 NP_001305451.1 2 - 3 211 87
NM_014410.4 NP_055225.1 2 - 3 250 87
NM_199167.1 NP_954636.1 2 - 3 176 87
XM_005258103.3 XP_005258160.1 4 - 5 400 87
XM_006722322.2 XP_006722385.1 3 - 4 368 87
XM_011525648.2 XP_011523950.1 4 - 5 443 87
XM_011525649.1 XP_011523951.1 4 - 5 443 87
XM_011525651.2 XP_011523953.1 3 - 4 1165 87
XM_011525652.1 XP_011523954.1 4 - 5 519 87
XM_011525653.2 XP_011523955.1 2 - 3 140 87
XM_017025707.1 XP_016881196.1 5 - 6 1905 87
GenBank mRNA AF395889.1 - 2 - 3 165 87
AK299319.1 - 4 - 5 470 87
AL712819.1 - 2 - 3 176 87
BC025381.2 - 2 - 3 250 87
D63813.1 - 2 - 3 156 87

View Full Product Details