The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022840.4 | NP_073751.3 | 7 - 8 | 1979 | 137 |
XM_005258133.2 | XP_005258190.1 | 7 - 8 | 1018 | 137 | |
GenBank mRNA | AK026670.1 | - | 5 - 6 | 780 | 137 |
AK290001.1 | - | 7 - 8 | 1900 | 137 | |
AL834296.1 | - | 4 - 5 | 505 | 137 | |
BC111020.1 | - | 7 - 8 | 1962 | 137 | |
BC136766.1 | - | 7 - 8 | 1725 | 137 | |
BC136767.1 | - | 7 - 8 | 1726 | 137 | |
BX647373.1 | - | 5 - 6 | 3486 | 137 |