Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLAMF6
Entrez Gene ID
114836
Gene Name
SLAM family member 6
Gene Aliases
CD352, KALI, KALIb, Ly108, NTB-A, NTBA, SF2000
Chromosome Location
Chr.1: 160485030 - 160523268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184714.1 NP_001171643.1 4 - 5 822 161
NM_001184715.1 NP_001171644.1 4 - 5 675 161
NM_001184716.1 NP_001171645.1 3 - 4 489 161
NM_052931.4 NP_443163.1 4 - 5 822 161
XM_017000215.1 XP_016855704.1 4 - 5 828 161
XM_017000216.1 XP_016855705.1 4 - 5 648 161
XM_017000217.1 XP_016855706.1 3 - 4 490 161
GenBank mRNA AJ277141.1 - 4 - 5 764 161
AJ306388.1 - 4 - 5 761 161
AK125624.1 - 4 - 5 822 161
AK300013.1 - 3 - 4 472 161
AK301026.1 - 3 - 4 479 161
AK303990.1 - 4 - 5 668 161
AK313549.1 - 4 - 5 774 161
AL832854.1 - 4 - 5 803 161
AY358159.1 - 4 - 5 813 161
BC113893.1 - 4 - 5 779 161
BC114495.1 - 4 - 5 779 161

View Full Product Details