Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLD3
Entrez Gene ID
23646
Gene Name
phospholipase D family member 3
Gene Aliases
AD19, HU-K4, HUK4
Chromosome Location
Chr.19: 40348395 - 40378490 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031696.3 NP_001026866.1 11 - 12 1583 70
NM_001291311.1 NP_001278240.1 11 - 12 1517 70
NM_012268.3 NP_036400.2 11 - 12 1807 70
XM_005258704.1 XP_005258761.1 11 - 12 1728 70
XM_005258707.4 XP_005258764.1 11 - 12 1510 70
XM_005258708.3 XP_005258765.1 10 - 11 1403 70
XM_005258709.4 XP_005258766.1 10 - 11 1391 70
XM_005258710.4 XP_005258767.1 10 - 11 1432 70
XM_006723122.1 XP_006723185.1 11 - 12 1895 70
XM_011526692.1 XP_011524994.1 11 - 12 1890 70
XM_011526693.1 XP_011524995.1 11 - 12 1610 70
XM_017026546.1 XP_016882035.1 12 - 13 1826 70
XM_017026547.1 XP_016882036.1 11 - 12 1394 70
XM_017026548.1 XP_016882037.1 11 - 12 1545 70
XM_017026549.1 XP_016882038.1 11 - 12 1648 70
GenBank mRNA BC000553.1 - 7 - 8 1053 70
BC036327.1 - 11 - 12 1308 70
BC096820.1 - 11 - 12 1506 70
U60644.1 - 11 - 12 1513 70

View Full Product Details