Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ETHE1
Entrez Gene ID
23474
Gene Name
ETHE1, persulfide dioxygenase
Gene Aliases
HSCO, YF13H12
Chromosome Location
Chr.19: 43506719 - 43527256 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320867.1 NP_001307796.1 5 - 6 642 80
NM_001320868.1 NP_001307797.1 4 - 5 526 80
NM_001320869.1 NP_001307798.1 3 - 4 381 80
NM_014297.4 NP_055112.2 5 - 6 675 80
XM_005258687.3 XP_005258744.1 5 - 6 772 80
GenBank mRNA AK309880.1 - 5 - 6 620 80
AK315346.1 - 5 - 6 638 80
BC008250.1 - 5 - 6 625 80
BG706505.1 - 4 - 5 475 80
D83198.2 - 5 - 6 664 80
KU178565.1 - 5 - 6 596 80
KU178566.1 - 5 - 6 563 80
KU178567.1 - 3 - 4 302 80

View Full Product Details