The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000702.3 | NP_000693.1 | 2 - 3 | 248 | 66 |
GenBank mRNA | AB018321.2 | - | 2 - 3 | 205 | 66 |
AK091617.1 | - | 2 - 3 | 219 | 66 | |
AK314296.1 | - | 2 - 3 | 219 | 66 | |
AL831991.1 | - | 1 - 2 | 37 | 66 | |
AL831997.1 | - | 1 - 2 | 63 | 66 | |
BC052271.1 | - | 2 - 3 | 199 | 66 | |
DA221602.1 | - | 2 - 3 | 248 | 66 | |
KU177927.1 | - | 1 - 2 | 116 | 66 | |
KU177928.1 | - | 1 - 2 | 116 | 66 |