The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000702.3 | NP_000693.1 | 4 - 5 | 509 | 83 |
GenBank mRNA | AB018321.2 | - | 4 - 5 | 466 | 83 |
AK091617.1 | - | 4 - 5 | 480 | 83 | |
AK314296.1 | - | 4 - 5 | 480 | 83 | |
AL831991.1 | - | 3 - 4 | 298 | 83 | |
AL831997.1 | - | 3 - 4 | 324 | 83 | |
AY946014.1 | - | 1 - 2 | 132 | 83 | |
BC052271.1 | - | 4 - 5 | 460 | 83 | |
KU177927.1 | - | 3 - 4 | 377 | 83 | |
KU177928.1 | - | 3 - 4 | 377 | 83 |