The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000702.3 | NP_000693.1 | 10 - 11 | 1463 | 67 |
GenBank mRNA | AB018321.2 | - | 10 - 11 | 1420 | 67 |
AK091617.1 | - | 10 - 11 | 1434 | 67 | |
AK124581.1 | - | 8 - 9 | 1135 | 67 | |
AK126573.1 | - | 3 - 4 | 463 | 67 | |
AK295729.1 | - | 3 - 4 | 440 | 67 | |
AK308773.1 | - | 2 - 3 | 570 | 67 | |
AK314296.1 | - | 10 - 11 | 1434 | 67 | |
AL831991.1 | - | 9 - 10 | 1252 | 67 | |
AL831997.1 | - | 9 - 10 | 1278 | 67 | |
BC052271.1 | - | 10 - 11 | 1414 | 67 | |
KU177927.1 | - | 9 - 10 | 1331 | 67 |