The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278547.1 | NP_001265476.1 | 7 - 8 | 912 | 87 |
NM_001323321.1 | NP_001310250.1 | 7 - 8 | 912 | 87 | |
NM_001323322.1 | NP_001310251.1 | 9 - 10 | 1175 | 87 | |
NM_001323323.1 | NP_001310252.1 | 8 - 9 | 981 | 87 | |
NM_001323325.1 | NP_001310254.1 | 8 - 9 | 1106 | 87 | |
NM_001323330.1 | NP_001310259.1 | 8 - 9 | 1106 | 87 | |
NM_139046.3 | NP_620634.1 | 9 - 10 | 1175 | 87 | |
NR_136585.1 | 7 - 8 | 912 | 87 | ||
GenBank mRNA | AB451271.1 | - | 6 - 7 | 680 | 87 |
AB451398.1 | - | 6 - 7 | 680 | 87 | |
AK292523.1 | - | 7 - 8 | 808 | 87 | |
BC130570.1 | - | 6 - 7 | 742 | 87 | |
BC130572.1 | - | 6 - 7 | 742 | 87 | |
BC144063.1 | - | 6 - 7 | 742 | 87 | |
U35004.1 | - | 6 - 7 | 698 | 87 | |
U35005.1 | - | 6 - 7 | 693 | 87 |