Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DLEC1
- Entrez Gene ID
-
9940
- Gene Name
- deleted in lung and esophageal cancer 1
- Gene Aliases
-
CFAP81, DLC-1, DLC1, F56
- Chromosome Location
-
Chr.3: 38038595 - 38122741 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY789464.1 |
- |
4 - 5 |
318 |
93 |
View Full Product Details