The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297717.1 | NP_001284646.1 | 3 - 4 | 675 | 82 |
NM_001297720.1 | NP_001284649.1 | 3 - 4 | 675 | 82 | |
NM_138740.3 | NP_620077.1 | 2 - 3 | 573 | 82 | |
GenBank mRNA | AJ243663.1 | - | 2 - 3 | 395 | 82 |
AL540989.3 | - | 3 - 4 | 462 | 82 | |
BC088353.1 | - | 3 - 4 | 495 | 82 | |
DR007097.1 | - | 2 - 3 | 455 | 82 |